The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005110.3 | NP_005101.1 | 17 - 18 | 2013 | 59 |
XM_017010100.1 | XP_016865589.1 | 16 - 17 | 1898 | 59 | |
GenBank mRNA | AB016789.1 | - | 17 - 18 | 1942 | 59 |
AK001242.1 | - | 7 - 8 | 895 | 59 | |
AK022507.1 | - | 17 - 18 | 1953 | 59 | |
AK297750.1 | - | 15 - 16 | 1649 | 59 | |
AK314551.1 | - | 17 - 18 | 1953 | 59 | |
BC000012.2 | - | 17 - 18 | 1951 | 59 | |
BT009818.1 | - | 16 - 17 | 1843 | 59 | |
KU178494.1 | - | 16 - 17 | 1843 | 59 |