The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037175.2 | NP_001032252.1 | 5 - 6 | 904 | 110 |
XM_017001589.1 | XP_016857078.1 | 6 - 7 | 1106 | 110 | |
GenBank mRNA | BC004888.2 | - | 5 - 6 | 785 | 110 |