Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP1
Entrez Gene ID
22861
Gene Name
NLR family pyrin domain containing 1
Gene Aliases
CARD7, CIDED, CLR17.1, DEFCAP, DEFCAP-L/S, NAC, NALP1, PP1044, SLEV1, VAMAS1
Chromosome Location
Chr.17: 5501399 - 5584512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033053.2 NP_001028225.1 3 - 4 1209 74
NM_014922.4 NP_055737.1 3 - 4 1209 74
NM_033004.3 NP_127497.1 3 - 4 1209 74
NM_033006.3 NP_127499.1 3 - 4 1209 74
NM_033007.3 NP_127500.1 3 - 4 1209 74
GenBank mRNA AB023143.1 - 3 - 4 1176 74
AF229059.1 - 3 - 4 654 74
AF229060.1 - 3 - 4 654 74
AF229061.1 - 3 - 4 654 74
AF229062.1 - 3 - 4 654 74
AF298548.1 - 4 - 5 813 74
AF310105.1 - 3 - 4 1172 74
AK310936.1 - 4 - 5 1020 74

View Full Product Details