Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP1
Entrez Gene ID
22861
Gene Name
NLR family pyrin domain containing 1
Gene Aliases
CARD7, CIDED, CLR17.1, DEFCAP, DEFCAP-L/S, NAC, NALP1, PP1044, SLEV1, VAMAS1
Chromosome Location
Chr.17: 5501399 - 5584512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033053.2 NP_001028225.1 4 - 5 2911 90
NM_014922.4 NP_055737.1 4 - 5 2911 90
NM_033004.3 NP_127497.1 4 - 5 2911 90
NM_033006.3 NP_127499.1 4 - 5 2911 90
NM_033007.3 NP_127500.1 4 - 5 2911 90
GenBank mRNA AB023143.1 - 4 - 5 2878 90
AF229059.1 - 4 - 5 2356 90
AF229060.1 - 4 - 5 2356 90
AF229061.1 - 4 - 5 2356 90
AF229062.1 - 4 - 5 2356 90
AF298548.1 - 5 - 6 90
AF310105.1 - 4 - 5 2874 90
AK026393.1 - 1 - 2 834 90
AK026398.1 - 1 - 2 173 90
AK310936.1 - 5 - 6 2722 90
AL117470.1 - 1 - 2 1514 90

View Full Product Details