The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033053.2 | NP_001028225.1 | 13 - 14 | 4349 | 121 |
NM_033004.3 | NP_127497.1 | 13 - 14 | 4337 | 121 | |
NM_033006.3 | NP_127499.1 | 12 - 13 | 4247 | 121 | |
GenBank mRNA | AF229060.1 | - | 13 - 14 | 3782 | 121 |
AF229062.1 | - | 12 - 13 | 3692 | 121 | |
AK026393.1 | - | 10 - 11 | 2261 | 121 | |
AK026398.1 | - | 10 - 11 | 1610 | 121 | |
AL117470.1 | - | 10 - 11 | 2940 | 121 |