Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC7A
Entrez Gene ID
57217
Gene Name
tetratricopeptide repeat domain 7A
Gene Aliases
GIDID, MINAT, TTC7
Chromosome Location
Chr.2: 46915931 - 47076136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288951.1 NP_001275880.1 18 - 19 2461 62
NM_001288953.1 NP_001275882.1 18 - 19 2380 62
NM_001288955.1 NP_001275884.1 16 - 17 2231 62
NM_020458.3 NP_065191.2 17 - 18 2389 62
XM_005264439.3 XP_005264496.1 18 - 19 2542 62
XM_011532998.2 XP_011531300.1 19 - 20 2607 62
XM_011533000.2 XP_011531302.1 14 - 15 1464 62
XM_011533001.2 XP_011531303.1 10 - 11 1254 62
XM_017004524.1 XP_016860013.1 16 - 17 2216 62
XM_017004525.1 XP_016860014.1 17 - 18 1874 62
XM_017004526.1 XP_016860015.1 14 - 15 2084 62
XM_017004527.1 XP_016860016.1 17 - 18 2484 62
XM_017004528.1 XP_016860017.1 18 - 19 2549 62
XM_017004529.1 XP_016860018.1 17 - 18 2332 62
GenBank mRNA AB032966.2 - 18 - 19 1969 62
AK056464.1 - 18 - 19 2352 62
AL834383.1 - 17 - 18 2389 62
BC001978.1 - 8 - 9 798 62
BC065554.1 - 14 - 15 1507 62
BC082965.1 - 9 - 10 1128 62
BC111487.1 - 18 - 19 2349 62
BC114365.2 - 16 - 17 1862 62

View Full Product Details