The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288951.1 | NP_001275880.1 | 20 - 21 | 2791 | 68 |
NM_001288953.1 | NP_001275882.1 | 20 - 21 | 2710 | 68 | |
NM_001288955.1 | NP_001275884.1 | 18 - 19 | 2561 | 68 | |
NM_020458.3 | NP_065191.2 | 19 - 20 | 2719 | 68 | |
XM_005264439.3 | XP_005264496.1 | 20 - 21 | 2872 | 68 | |
XM_011532998.2 | XP_011531300.1 | 21 - 22 | 2937 | 68 | |
XM_011533000.2 | XP_011531302.1 | 16 - 17 | 1794 | 68 | |
XM_011533001.2 | XP_011531303.1 | 12 - 13 | 1584 | 68 | |
XM_017004524.1 | XP_016860013.1 | 18 - 19 | 2546 | 68 | |
XM_017004525.1 | XP_016860014.1 | 19 - 20 | 2204 | 68 | |
XM_017004526.1 | XP_016860015.1 | 16 - 17 | 2414 | 68 | |
XM_017004527.1 | XP_016860016.1 | 19 - 20 | 2814 | 68 | |
XM_017004528.1 | XP_016860017.1 | 20 - 21 | 2879 | 68 | |
GenBank mRNA | AB032966.2 | - | 20 - 21 | 2299 | 68 |
AK056464.1 | - | 20 - 21 | 2682 | 68 | |
AL834383.1 | - | 19 - 20 | 2719 | 68 | |
BC001978.1 | - | 10 - 11 | 1128 | 68 | |
BC027457.2 | - | 2 - 3 | 281 | 68 | |
BC065554.1 | - | 16 - 17 | 1837 | 68 | |
BC082965.1 | - | 11 - 12 | 1458 | 68 | |
BC111487.1 | - | 20 - 21 | 2679 | 68 | |
BC114365.2 | - | 18 - 19 | 2192 | 68 |