Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS16
Entrez Gene ID
170690
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 16
Gene Aliases
ADAMTS16s
Chromosome Location
Chr.5: 5140330 - 5320304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139056.3 NP_620687.2 20 - 21 3325 133
NR_136935.1 19 - 20 3194 133
GenBank mRNA AB095949.1 - 17 - 18 2580 133

View Full Product Details