The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159597.2 | NP_001153069.1 | 7 - 8 | 814 | 78 |
NM_001282682.1 | NP_001269611.1 | 9 - 10 | 919 | 78 | |
NM_001282687.1 | NP_001269616.1 | 9 - 10 | 968 | 78 | |
NM_015677.3 | NP_056492.2 | 7 - 8 | 814 | 78 | |
NR_104223.1 | 9 - 10 | 2373 | 78 | ||
NR_104225.1 | 7 - 8 | 1009 | 78 | ||
NR_104226.1 | 9 - 10 | 1534 | 78 | ||
GenBank mRNA | AK026507.1 | - | 7 - 8 | 749 | 78 |
AK026586.1 | - | 7 - 8 | 1009 | 78 | |
AK123829.1 | - | 10 - 11 | 2612 | 78 | |
AK292312.1 | - | 9 - 10 | 863 | 78 | |
AL050373.1 | - | 9 - 10 | 981 | 78 | |
BC008374.1 | - | 7 - 8 | 723 | 78 | |
BC008375.1 | - | 7 - 8 | 741 | 78 | |
BC043234.1 | - | 9 - 10 | 2378 | 78 | |
CR457361.1 | - | 6 - 7 | 701 | 78 |