Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC17
Entrez Gene ID
55761
Gene Name
tetratricopeptide repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.11: 43358885 - 43494933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307943.1 NP_001294872.1 19 - 20 2902 61
GenBank mRNA AF161431.1 - 3 - 4 502 61
AK001540.1 - 14 - 15 2137 61
AK001752.1 - 19 - 20 2855 61
BC033000.1 - 19 - 20 2848 61
BX640972.1 - 14 - 15 2742 61

View Full Product Details