Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AGBL2
- Entrez Gene ID
-
79841
- Gene Name
- ATP/GTP binding protein like 2
- Gene Aliases
-
CCP2
- Chromosome Location
-
Chr.11: 47659168 - 47715397 on Build GRCh38
View Full Product Details