The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138431.2 | NP_612440.1 | 2 - 3 | 1128 | 148 |
XM_011516806.2 | XP_011515108.1 | 2 - 3 | 2075 | 148 | |
XM_017013005.1 | XP_016868494.1 | 2 - 3 | 2083 | 148 | |
GenBank mRNA | BC011982.1 | - | 2 - 3 | 995 | 148 |