Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGL1
Entrez Gene ID
2267
Gene Name
fibrinogen like 1
Gene Aliases
HFREP1, HP-041, LFIRE-1, LFIRE1
Chromosome Location
Chr.8: 17864380 - 17909983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004467.3 NP_004458.3 5 - 6 608 171
NM_147203.2 NP_671736.2 5 - 6 696 171
NM_201552.1 NP_963846.1 6 - 7 735 171
NM_201553.1 NP_963847.1 6 - 7 823 171
XM_017013232.1 XP_016868721.1 6 - 7 1585 171
XM_017013233.1 XP_016868722.1 4 - 5 660 171
GenBank mRNA AF042354.1 - 5 - 6 644 171
AF168954.1 - 5 - 6 631 171
AK225616.1 - 5 - 6 594 171
AK225619.1 - 5 - 6 583 171
AK289448.1 - 5 - 6 594 171
AL514837.3 - 3 - 4 171
AX772838.1 - 6 - 7 688 171
BC007047.1 - 5 - 6 604 171
BT006635.1 - 4 - 5 499 171
D14446.1 - 5 - 6 577 171
D87342.1 - 4 - 5 504 171

View Full Product Details