Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKS6
Entrez Gene ID
203286
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
ANKRD14, NPHP16, PKDR1, SAMD6
Chromosome Location
Chr.9: 98732009 - 98796542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011518357.2 XP_011516659.1 1 - 1 1471 66
XM_017014446.1 XP_016869935.1 1 - 1 1471 66
GenBank mRNA BC012981.2 - 1 - 1 486 66

View Full Product Details