The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135155.2 | NP_001128627.1 | 1 - 2 | 141 | 63 |
NM_001289978.1 | NP_001276907.1 | 1 - 2 | 141 | 63 | |
NM_004647.3 | NP_004638.2 | 1 - 2 | 141 | 63 | |
XM_005259289.2 | XP_005259346.1 | 1 - 2 | 141 | 63 | |
XM_006723409.2 | XP_006723472.1 | 1 - 2 | 141 | 63 | |
XM_011527356.1 | XP_011525658.1 | 1 - 2 | 141 | 63 | |
GenBank mRNA | AK308563.1 | - | 1 - 2 | 141 | 63 |