Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOTCH2
Entrez Gene ID
4853
Gene Name
notch 2
Gene Aliases
AGS2, HJCYS, hN2
Chromosome Location
Chr.1: 119911553 - 120073449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001200001.1 NP_001186930.1 20 - 21 3638 73
NM_024408.3 NP_077719.2 20 - 21 3638 73
XM_005270901.3 XP_005270958.1 21 - 22 5952 73
XM_011541519.2 XP_011539821.1 20 - 21 8746 73
XM_011541520.2 XP_011539822.1 20 - 21 5656 73
XM_017001372.1 XP_016856861.1 20 - 21 3308 73
XM_017001373.1 XP_016856862.1 20 - 21 3287 73
GenBank mRNA AF308601.1 - 20 - 21 3353 73
AF315356.1 - 20 - 21 3353 73
BC071562.1 - 20 - 21 3587 73

View Full Product Details