The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024408.3 | NP_077719.2 | 28 - 29 | 5511 | 88 |
XM_005270901.3 | XP_005270958.1 | 29 - 30 | 7825 | 88 | |
XM_011541519.2 | XP_011539821.1 | 28 - 29 | 10619 | 88 | |
XM_011541520.2 | XP_011539822.1 | 28 - 29 | 7529 | 88 | |
XM_017001372.1 | XP_016856861.1 | 28 - 29 | 5181 | 88 | |
XM_017001373.1 | XP_016856862.1 | 28 - 29 | 5160 | 88 | |
GenBank mRNA | AF308601.1 | - | 28 - 29 | 5226 | 88 |
AF315356.1 | - | 28 - 29 | 5226 | 88 | |
AK027869.1 | - | 2 - 3 | 350 | 88 |