The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001200001.1 | NP_001186930.1 | 13 - 14 | 2512 | 65 |
NM_024408.3 | NP_077719.2 | 13 - 14 | 2512 | 65 | |
XM_005270901.3 | XP_005270958.1 | 14 - 15 | 4826 | 65 | |
XM_011541519.2 | XP_011539821.1 | 13 - 14 | 7620 | 65 | |
XM_011541520.2 | XP_011539822.1 | 13 - 14 | 4530 | 65 | |
XM_017001372.1 | XP_016856861.1 | 13 - 14 | 2182 | 65 | |
XM_017001373.1 | XP_016856862.1 | 13 - 14 | 2161 | 65 | |
GenBank mRNA | AF308601.1 | - | 13 - 14 | 2227 | 65 |
AF315356.1 | - | 13 - 14 | 2227 | 65 | |
AK307843.1 | - | 12 - 13 | 2353 | 65 | |
BC071562.1 | - | 13 - 14 | 2461 | 65 | |
GQ231534.1 | - | 13 - 14 | 2374 | 65 |