The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320044.1 | NP_001306973.1 | 10 - 11 | 1710 | 76 |
NM_012235.3 | NP_036367.2 | 13 - 14 | 2195 | 76 | |
XM_005264967.1 | XP_005265024.1 | 13 - 14 | 2122 | 76 | |
XM_005264968.1 | XP_005265025.1 | 10 - 11 | 1864 | 76 | |
XM_005264971.1 | XP_005265028.1 | 7 - 8 | 1275 | 76 | |
XM_011533501.1 | XP_011531803.1 | 13 - 14 | 2222 | 76 | |
XM_011533502.2 | XP_011531804.1 | 5 - 6 | 871 | 76 | |
XM_017005918.1 | XP_016861407.1 | 13 - 14 | 2209 | 76 | |
XM_017005919.1 | XP_016861408.1 | 13 - 14 | 2142 | 76 | |
XM_017005920.1 | XP_016861409.1 | 13 - 14 | 2142 | 76 | |
XM_017005921.1 | XP_016861410.1 | 7 - 8 | 1275 | 76 | |
GenBank mRNA | AK027402.1 | - | 7 - 8 | 1188 | 76 |
AK291484.1 | - | 8 - 9 | 1388 | 76 | |
AK300311.1 | - | 10 - 11 | 1600 | 76 | |
BC020987.1 | - | 5 - 6 | 916 | 76 | |
D83782.2 | - | 13 - 14 | 2181 | 76 |