The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287489.1 | NP_001274418.1 | 38 - 39 | 4922 | 123 |
NM_004802.3 | NP_004793.2 | 21 - 22 | 2720 | 123 | |
NM_194248.2 | NP_919224.1 | 38 - 39 | 4922 | 123 | |
NM_194322.2 | NP_919303.1 | 20 - 21 | 2874 | 123 | |
NM_194323.2 | NP_919304.1 | 21 - 22 | 2720 | 123 | |
XM_017005338.1 | XP_016860827.1 | 38 - 39 | 4735 | 123 | |
GenBank mRNA | AF107403.1 | - | 21 - 22 | 2720 | 123 |
AF183185.1 | - | 38 - 39 | 4922 | 123 | |
AF183186.1 | - | 20 - 21 | 2874 | 123 | |
AF183187.1 | - | 21 - 22 | 2720 | 123 | |
AK296272.1 | - | 11 - 12 | 1277 | 123 |