The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287489.1 | NP_001274418.1 | 42 - 43 | 5417 | 95 |
NM_004802.3 | NP_004793.2 | 25 - 26 | 3215 | 95 | |
NM_194248.2 | NP_919224.1 | 42 - 43 | 5417 | 95 | |
NM_194322.2 | NP_919303.1 | 24 - 25 | 3369 | 95 | |
NM_194323.2 | NP_919304.1 | 25 - 26 | 3215 | 95 | |
XM_017005338.1 | XP_016860827.1 | 42 - 43 | 5230 | 95 | |
GenBank mRNA | AF107403.1 | - | 25 - 26 | 3215 | 95 |
AF183185.1 | - | 42 - 43 | 5417 | 95 | |
AF183186.1 | - | 24 - 25 | 3369 | 95 | |
AF183187.1 | - | 25 - 26 | 3215 | 95 | |
AK296272.1 | - | 15 - 16 | 1772 | 95 |