The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004802.3 | NP_004793.2 | 28 - 29 | 3741 | 83 |
NM_194248.2 | NP_919224.1 | 45 - 46 | 5943 | 83 | |
NM_194322.2 | NP_919303.1 | 27 - 28 | 3895 | 83 | |
XM_017005338.1 | XP_016860827.1 | 45 - 46 | 5756 | 83 | |
GenBank mRNA | AF107403.1 | - | 28 - 29 | 3741 | 83 |
AF183185.1 | - | 45 - 46 | 5943 | 83 | |
AF183186.1 | - | 27 - 28 | 3895 | 83 |