The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197320.1 | NP_001184249.1 | 3 - 4 | 682 | 89 |
NM_021960.4 | NP_068779.1 | 2 - 3 | 1141 | 89 | |
GenBank mRNA | AF118124.1 | - | 2 - 3 | 982 | 89 |
AK294462.1 | - | 3 - 4 | 713 | 89 | |
AK297217.1 | - | 3 - 4 | 677 | 89 | |
AK300499.1 | - | 3 - 4 | 787 | 89 | |
AK304775.1 | - | 3 - 4 | 823 | 89 | |
AK312508.1 | - | 2 - 3 | 1013 | 89 | |
AK316267.1 | - | 3 - 4 | 969 | 89 | |
BC017197.2 | - | 2 - 3 | 987 | 89 | |
BC071897.1 | - | 2 - 3 | 989 | 89 | |
BC107735.1 | - | 2 - 3 | 1004 | 89 | |
BT006640.1 | - | 2 - 3 | 933 | 89 | |
FJ917536.1 | - | 3 - 4 | 474 | 89 | |
L08246.1 | - | 2 - 3 | 993 | 89 |