Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INSRR
Entrez Gene ID
3645
Gene Name
insulin receptor related receptor
Gene Aliases
IRR
Chromosome Location
Chr.1: 156840873 - 156858920 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014215.2 NP_055030.1 5 - 6 1529 90
GenBank mRNA J05046.1 - 4 - 5 1145 90

View Full Product Details