Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIRE1
Entrez Gene ID
56907
Gene Name
spire type actin nucleation factor 1
Gene Aliases
Spir-1
Chromosome Location
Chr.18: 12446512 - 12658102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005258122.4 XP_005258179.1 12 - 13 2003 69
XM_011525701.1 XP_011524003.1 13 - 14 2051 69
XM_011525702.1 XP_011524004.1 13 - 14 1563 69

View Full Product Details