Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR6
Entrez Gene ID
63897
Gene Name
HEAT repeat containing 6
Gene Aliases
ABC1
Chromosome Location
Chr.17: 60041363 - 60078931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022070.4 NP_071353.4 7 - 8 950 93
XM_005257570.3 XP_005257627.1 8 - 9 1138 93
XM_011525110.2 XP_011523412.1 6 - 7 837 93
XM_011525111.2 XP_011523413.1 6 - 7 799 93
XM_017024936.1 XP_016880425.1 7 - 8 1211 93
XM_017024937.1 XP_016880426.1 7 - 8 2887 93
XM_017024938.1 XP_016880427.1 7 - 8 940 93
XM_017024939.1 XP_016880428.1 7 - 8 1211 93
GenBank mRNA AF349752.1 - 8 - 9 895 93
AK127708.1 - 7 - 8 944 93
AK314655.1 - 7 - 8 940 93
BC119771.1 - 7 - 8 940 93
BC119813.1 - 7 - 8 941 93
BX640819.1 - 7 - 8 941 93
BX640831.1 - 7 - 8 941 93
CR627418.1 - 7 - 8 940 93

View Full Product Details