The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320424.1 | NP_001307353.1 | 5 - 6 | 644 | 66 |
NM_152328.4 | NP_689541.1 | 5 - 6 | 557 | 66 | |
NM_199165.2 | NP_954634.1 | 5 - 6 | 661 | 66 | |
XM_006720026.3 | XP_006720089.1 | 6 - 7 | 556 | 66 | |
XM_011536412.1 | XP_011534714.1 | 6 - 7 | 630 | 66 | |
XM_011536414.2 | XP_011534716.1 | 6 - 7 | 313 | 66 | |
GenBank mRNA | AK095921.1 | - | 5 - 6 | 553 | 66 |
AK096124.1 | - | 3 - 4 | 1404 | 66 | |
AK096720.1 | - | 5 - 6 | 595 | 66 | |
AY037159.1 | - | 5 - 6 | 542 | 66 | |
BC032039.1 | - | 5 - 6 | 618 | 66 | |
BC047904.1 | - | 5 - 6 | 557 | 66 | |
BX248286.1 | - | 5 - 6 | 514 | 66 |