The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252335.1 | NP_001239264.1 | 12 - 13 | 3068 | 64 |
NM_032866.4 | NP_116255.2 | 11 - 12 | 2953 | 64 | |
XM_005254726.3 | XP_005254783.1 | 11 - 12 | 2971 | 64 | |
XM_005254727.4 | XP_005254784.1 | 12 - 13 | 3349 | 64 | |
XM_011522120.1 | XP_011520422.1 | 12 - 13 | 3007 | 64 | |
XM_011522121.1 | XP_011520423.1 | 11 - 12 | 3130 | 64 | |
XM_017022685.1 | XP_016878174.1 | 11 - 12 | 3588 | 64 | |
XM_017022686.1 | XP_016878175.1 | 11 - 12 | 3586 | 64 | |
GenBank mRNA | AB051536.2 | - | 10 - 11 | 2112 | 64 |
AK027863.1 | - | 6 - 7 | 863 | 64 | |
AK307410.1 | - | 11 - 12 | 2953 | 64 | |
AY274808.1 | - | 11 - 12 | 2951 | 64 | |
AY610514.1 | - | 12 - 13 | 3048 | 64 | |
BC093827.1 | - | 10 - 11 | 2895 | 64 | |
BC112049.1 | - | 10 - 11 | 2907 | 64 | |
BC118918.1 | - | 10 - 11 | 2894 | 64 |