Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGNL1
Entrez Gene ID
84952
Gene Name
cingulin like 1
Gene Aliases
JACOP, PCING
Chromosome Location
Chr.15: 57376487 - 57550727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252335.1 NP_001239264.1 4 - 5 1893 128
NM_032866.4 NP_116255.2 3 - 4 1778 128
XM_005254726.3 XP_005254783.1 3 - 4 1796 128
XM_005254727.4 XP_005254784.1 4 - 5 2174 128
XM_011522120.1 XP_011520422.1 4 - 5 1832 128
XM_011522121.1 XP_011520423.1 3 - 4 1955 128
XM_017022685.1 XP_016878174.1 3 - 4 2413 128
XM_017022686.1 XP_016878175.1 3 - 4 2411 128
GenBank mRNA AB051536.2 - 2 - 3 937 128
AK056673.1 - 3 - 4 1778 128
AK307410.1 - 3 - 4 1778 128
AY274808.1 - 3 - 4 1776 128
AY610514.1 - 4 - 5 1873 128
BC030995.1 - 3 - 4 1776 128
BC062770.1 - 2 - 3 1168 128
BC093827.1 - 2 - 3 1720 128
BC112049.1 - 2 - 3 1732 128
BC118918.1 - 2 - 3 1719 128

View Full Product Details