Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTPBP2
Entrez Gene ID
54676
Gene Name
GTP binding protein 2
Gene Aliases
-
Chromosome Location
Chr.6: 43620481 - 43629246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286216.1 NP_001273145.1 7 - 8 983 96
NM_019096.4 NP_061969.3 7 - 8 1178 96
XM_017010976.1 XP_016866465.1 7 - 8 1172 96
GenBank mRNA AB024574.1 - 8 - 9 1021 96
AF168990.1 - 7 - 8 975 96
AJ420518.1 - 2 - 3 341 96
AK290267.1 - 7 - 8 983 96
AL834331.1 - 7 - 8 960 96
BC020980.2 - 3 - 4 594 96
BC028347.2 - 5 - 6 886 96
BC064968.1 - 7 - 8 1131 96

View Full Product Details