Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C19orf25
Entrez Gene ID
148223
Gene Name
chromosome 19 open reading frame 25
Gene Aliases
-
Chromosome Location
Chr.19: 1473201 - 1479552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152482.2 NP_689695.2 1 - 2 67 56
XM_017026374.1 XP_016881863.1 1 - 2 390 56
GenBank mRNA AK075267.1 - 1 - 2 50 56
AK093985.1 - 1 - 2 32 56
AK297944.1 - 1 - 2 54 56
AL122089.1 - 1 - 2 38 56
BC018441.1 - 1 - 2 37 56
BP210473.1 - 1 - 2 67 56

View Full Product Details