Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFR
Entrez Gene ID
51663
Gene Name
zinc finger RNA binding protein
Gene Aliases
SPG71, ZFR1
Chromosome Location
Chr.5: 32354350 - 32444738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016107.3 NP_057191.2 8 - 9 1591 85
GenBank mRNA AJ314790.1 - 8 - 9 1544 85
AK292214.1 - 8 - 9 1591 85
BC030540.1 - 8 - 9 1563 85
BC051893.1 - 1 - 2 182 85
BC062986.1 - 1 - 2 107 85
BC137084.1 - 8 - 9 1555 85
BC137085.1 - 8 - 9 1555 85

View Full Product Details