Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RIMKLB
- Entrez Gene ID
-
57494
- Gene Name
- ribosomal modification protein rimK like family member B
- Gene Aliases
-
FAM80B, NAAGS, NAAGS-I
- Chromosome Location
-
Chr.12: 8681606 - 8783098 on Build GRCh38
View Full Product Details