Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUSP9
Entrez Gene ID
1852
Gene Name
dual specificity phosphatase 9
Gene Aliases
MKP-4, MKP4
Chromosome Location
Chr.X: 153642443 - 153651326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011531124.1 XP_011529426.1 1 - 2 217 58

View Full Product Details