The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018164.2 | NP_060634.2 | 13 - 14 | 2109 | 100 |
XM_005253422.3 | XP_005253479.1 | 12 - 13 | 1897 | 100 | |
XM_011520750.2 | XP_011519052.1 | 13 - 14 | 1660 | 100 | |
XM_011520751.2 | XP_011519053.1 | 12 - 13 | 1915 | 100 | |
XM_011520752.2 | XP_011519054.1 | 12 - 13 | 1861 | 100 | |
XM_011520753.2 | XP_011519055.1 | 7 - 8 | 832 | 100 | |
XM_017019632.1 | XP_016875121.1 | 13 - 14 | 1642 | 100 | |
XM_017019633.1 | XP_016875122.1 | 12 - 13 | 2052 | 100 | |
XM_017019634.1 | XP_016875123.1 | 12 - 13 | 2034 | 100 | |
XM_017019635.1 | XP_016875124.1 | 12 - 13 | 1471 | 100 | |
XM_017019636.1 | XP_016875125.1 | 12 - 13 | 1843 | 100 | |
XM_017019637.1 | XP_016875126.1 | 11 - 12 | 1272 | 100 | |
GenBank mRNA | AF274950.1 | - | 4 - 5 | 521 | 100 |
AK001222.1 | - | 3 - 4 | 424 | 100 | |
AK001499.1 | - | 13 - 14 | 1774 | 100 | |
AK225336.1 | - | 13 - 14 | 1760 | 100 | |
AK297948.1 | - | 11 - 12 | 1449 | 100 | |
BC003081.2 | - | 13 - 14 | 1798 | 100 | |
BC008368.1 | - | 13 - 14 | 1756 | 100 |