Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASUN
Entrez Gene ID
55726
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
C12orf11, GCT1, Mat89Bb, NET48, SPATA30
Chromosome Location
Chr.12: 26905171 - 26938326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018164.2 NP_060634.2 6 - 7 1213 72
XM_005253422.3 XP_005253479.1 6 - 7 1172 72
XM_011520750.2 XP_011519052.1 6 - 7 746 72
XM_011520751.2 XP_011519053.1 6 - 7 1172 72
XM_011520752.2 XP_011519054.1 6 - 7 1172 72
XM_017019632.1 XP_016875121.1 6 - 7 746 72
XM_017019633.1 XP_016875122.1 5 - 6 1138 72
XM_017019634.1 XP_016875123.1 5 - 6 1138 72
XM_017019635.1 XP_016875124.1 6 - 7 746 72
XM_017019636.1 XP_016875125.1 6 - 7 1172 72
XM_017019637.1 XP_016875126.1 3 - 4 305 72
GenBank mRNA AK001492.1 - 6 - 7 842 72
AK001499.1 - 6 - 7 878 72
AK225336.1 - 6 - 7 864 72
AK297948.1 - 4 - 5 553 72
BC003081.2 - 6 - 7 902 72
BC008368.1 - 6 - 7 860 72

View Full Product Details