The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291593.1 | NP_001278522.1 | 21 - 22 | 3432 | 88 |
NM_001291594.1 | NP_001278523.1 | 20 - 21 | 3255 | 88 | |
NM_015102.4 | NP_055917.1 | 24 - 25 | 3742 | 88 | |
NR_111987.1 | 27 - 28 | 4289 | 88 | ||
XM_006710563.3 | XP_006710626.1 | 24 - 25 | 3588 | 88 | |
XM_011541213.1 | XP_011539515.1 | 23 - 24 | 3509 | 88 | |
XM_011541214.1 | XP_011539516.1 | 23 - 24 | 3470 | 88 | |
XM_011541215.1 | XP_011539517.1 | 22 - 23 | 3401 | 88 | |
XM_011541216.2 | XP_011539518.1 | 25 - 26 | 4216 | 88 | |
XM_011541217.2 | XP_011539519.1 | 24 - 25 | 4080 | 88 | |
XM_011541218.2 | XP_011539520.1 | 25 - 26 | 3749 | 88 | |
XM_017000996.1 | XP_016856485.1 | 23 - 24 | 3467 | 88 | |
XM_017000997.1 | XP_016856486.1 | 24 - 25 | 4011 | 88 | |
XM_017000998.1 | XP_016856487.1 | 23 - 24 | 3512 | 88 | |
XM_017000999.1 | XP_016856488.1 | 24 - 25 | 3507 | 88 | |
XM_017001000.1 | XP_016856489.1 | 23 - 24 | 3340 | 88 | |
XM_017001001.1 | XP_016856490.1 | 18 - 19 | 2779 | 88 | |
XM_017001003.1 | XP_016856492.1 | 16 - 17 | 2570 | 88 | |
GenBank mRNA | AB014573.1 | - | 19 - 20 | 2842 | 88 |
AF537130.1 | - | 24 - 25 | 3740 | 88 | |
AK124074.1 | - | 20 - 21 | 3208 | 88 | |
AK127333.1 | - | 1 - 2 | 638 | 88 | |
AK314326.1 | - | 21 - 22 | 3406 | 88 | |
AY118228.1 | - | 24 - 25 | 3677 | 88 | |
BC040520.1 | - | 27 - 28 | 4290 | 88 |