The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282927.1 | NP_001269856.1 | 1 - 2 | 159 | 71 |
XM_006712177.2 | XP_006712240.1 | 2 - 3 | 924 | 71 | |
XM_006712185.1 | XP_006712248.1 | 2 - 3 | 924 | 71 | |
XM_017003128.1 | XP_016858617.1 | 2 - 3 | 924 | 71 | |
XM_017003133.1 | XP_016858622.1 | 2 - 3 | 924 | 71 | |
XM_017003137.1 | XP_016858626.1 | 2 - 3 | 924 | 71 | |
XM_017003139.1 | XP_016858628.1 | 2 - 3 | 924 | 71 | |
XM_017003140.1 | XP_016858629.1 | 2 - 3 | 924 | 71 | |
XM_017003143.1 | XP_016858632.1 | 2 - 3 | 924 | 71 | |
GenBank mRNA | U23435.1 | - | 1 - 2 | 120 | 71 |