Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
Chromosome Location
Chr.22: 41699503 - 41799456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152513.3 NP_689726.3 26 - 27 3373 57
XM_011529935.1 XP_011528237.1 27 - 28 3391 57
XM_011529936.1 XP_011528238.1 26 - 27 3367 57
XM_011529937.1 XP_011528239.1 25 - 26 3322 57
XM_011529938.1 XP_011528240.1 25 - 26 3310 57
XM_011529939.1 XP_011528241.1 26 - 27 3427 57
XM_011529940.1 XP_011528242.1 25 - 26 3277 57
XM_011529941.1 XP_011528243.1 26 - 27 3382 57
XM_011529942.2 XP_011528244.1 26 - 27 3618 57
XM_011529943.1 XP_011528245.1 25 - 26 3151 57
XM_011529945.2 XP_011528247.1 26 - 27 3429 57
XM_011529946.1 XP_011528248.1 23 - 24 2995 57
XM_011529947.1 XP_011528249.1 21 - 22 2905 57
XM_011529948.2 XP_011528250.1 22 - 23 2799 57
XM_011529949.1 XP_011528251.1 21 - 22 2736 57
XM_011529952.1 XP_011528254.1 21 - 22 2873 57
XM_011529954.1 XP_011528256.1 16 - 17 2177 57
XM_017028633.1 XP_016884122.1 26 - 27 3322 57
XM_017028635.1 XP_016884124.1 12 - 13 1757 57
GenBank mRNA AK092934.1 - 12 - 13 1742 57
AK093341.1 - 6 - 7 878 57
AK094461.1 - 8 - 9 1159 57
AK097834.1 - 6 - 7 924 57
AK301391.1 - 3 - 4 692 57
AK301435.1 - 11 - 12 1579 57
AY952376.1 - 11 - 12 2085 57
AY952377.1 - 10 - 11 1980 57
BC032248.1 - 3 - 4 707 57
BC070111.1 - 22 - 23 3023 57

View Full Product Details