Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHDC2
Entrez Gene ID
23588
Gene Name
kelch domain containing 2
Gene Aliases
HCLP-1, HCLP1, LCP
Chromosome Location
Chr.14: 49767589 - 49785706 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014315.2 NP_055130.1 1 - 2 558 109
XM_006720094.3 XP_006720157.1 1 - 2 1038 109
GenBank mRNA AF113131.1 - 1 - 2 416 109
AF244137.1 - 1 - 2 493 109
AK001771.1 - 1 - 2 474 109
AK056298.1 - 1 - 2 1019 109
BC002335.2 - 1 - 2 485 109
BC024192.1 - 1 - 2 486 109
BX161429.1 - 1 - 2 453 109
CF994676.1 - 1 - 2 501 109
CR457205.1 - 1 - 2 158 109

View Full Product Details