The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111038.1 | NP_001104508.1 | 3 - 4 | 318 | 104 |
NM_001287150.1 | NP_001274079.1 | 3 - 4 | 318 | 104 | |
NM_145032.3 | NP_659469.3 | 3 - 4 | 495 | 104 | |
NR_105043.1 | 4 - 5 | 409 | 104 | ||
XM_005250205.3 | XP_005250262.1 | 4 - 5 | 401 | 104 | |
XM_005250207.3 | XP_005250264.1 | 4 - 5 | 401 | 104 | |
XM_005250208.3 | XP_005250265.1 | 4 - 5 | 401 | 104 | |
XM_005250209.2 | XP_005250266.1 | 3 - 4 | 299 | 104 | |
XM_011515928.2 | XP_011514230.1 | 4 - 5 | 401 | 104 | |
XM_011515929.2 | XP_011514231.1 | 4 - 5 | 401 | 104 | |
XM_011515930.2 | XP_011514232.1 | 3 - 4 | 365 | 104 | |
XM_011515932.2 | XP_011514234.1 | 4 - 5 | 401 | 104 | |
XM_017011850.1 | XP_016867339.1 | 4 - 5 | 345 | 104 | |
XM_017011851.1 | XP_016867340.1 | 4 - 5 | 401 | 104 | |
XM_017011852.1 | XP_016867341.1 | 4 - 5 | 551 | 104 | |
XM_017011853.1 | XP_016867342.1 | 3 - 4 | 351 | 104 | |
GenBank mRNA | AK097537.1 | - | 3 - 4 | 495 | 104 |
AY359238.1 | - | 3 - 4 | 202 | 104 | |
AY359239.1 | - | 3 - 4 | 202 | 104 | |
BC026121.1 | - | 4 - 5 | 412 | 104 | |
BC031285.1 | - | 3 - 4 | 310 | 104 | |
BG717734.1 | - | 4 - 5 | 414 | 104 |