Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
Chromosome Location
Chr.7: 140719331 - 140924764 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017012558.1 XP_016868047.1 9 - 10 1185 117
XM_017012559.1 XP_016868048.1 9 - 10 1185 117

View Full Product Details