The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304968.1 | NP_001291897.1 | 2 - 3 | 707 | 148 |
GenBank mRNA | AK093329.1 | - | 2 - 3 | 706 | 148 |