The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000192.3 | NP_000183.2 | 1 - 2 | 623 | 60 |
GenBank mRNA | AF221714.1 | - | 1 - 2 | 623 | 60 |
DA863182.1 | - | 1 - 2 | 227 | 60 | |
Y09445.1 | - | 1 - 2 | 623 | 60 |