Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS27
Entrez Gene ID
23107
Gene Name
mitochondrial ribosomal protein S27
Gene Aliases
MRP-S27, S27mt
Chromosome Location
Chr.5: 72219409 - 72320257 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286748.1 NP_001273677.1 2 - 3 186 137
NM_001286751.1 NP_001273680.1 2 - 3 154 137
NM_015084.2 NP_055899.2 2 - 3 186 137
GenBank mRNA AK025417.1 - 2 - 3 169 137
AK299412.1 - 2 - 3 160 137
AK300111.1 - 2 - 3 152 137
AK309065.1 - 2 - 3 136 137
BC064902.1 - 2 - 3 157 137
D87453.1 - 2 - 3 155 137
DC341760.1 - 2 - 3 186 137

View Full Product Details