Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCHIP1
Entrez Gene ID
29970
Gene Name
schwannomin interacting protein 1
Gene Aliases
SCHIP-1
Chromosome Location
Chr.3: 159273247 - 159897366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197107.1 NP_001184036.1 6 - 7 1848 72
NM_001197108.1 NP_001184037.1 5 - 6 1191 72
NM_001197109.1 NP_001184038.1 5 - 6 908 72
NM_014575.3 NP_055390.1 6 - 7 1887 72
GenBank mRNA AF070614.1 - 5 - 6 997 72
AF145713.1 - 6 - 7 1382 72
AF145714.1 - 5 - 6 686 72
AF145715.1 - 6 - 7 1343 72
AK091871.1 - 5 - 6 829 72
AK309223.1 - 6 - 7 1295 72
AK314662.1 - 6 - 7 1830 72
BC005947.1 - 5 - 6 724 72
BC036535.2 - 6 - 7 1887 72
BX648179.1 - 3 - 4 3319 72

Gene Symbol
IQCJ-SCHIP1
Entrez Gene ID
100505385
Gene Name
IQCJ-SCHIP1 readthrough
Gene Aliases
-
Chromosome Location
Chr.3: 159069252 - 159897366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197113.1 NP_001184042.1 9 - 10 1722 72
NM_001197114.1 NP_001184043.1 8 - 9 1641 72
GenBank mRNA AK096479.1 - 9 - 10 1628 72
DQ157847.1 - 7 - 8 1460 72
DQ157848.1 - 8 - 9 1541 72

View Full Product Details