Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCHIP1
Entrez Gene ID
29970
Gene Name
schwannomin interacting protein 1
Gene Aliases
SCHIP-1
Chromosome Location
Chr.3: 159273247 - 159897366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197107.1 NP_001184036.1 7 - 8 1943 60
NM_001197108.1 NP_001184037.1 6 - 7 1286 60
NM_001197109.1 NP_001184038.1 6 - 7 1003 60
NM_014575.3 NP_055390.1 7 - 8 1982 60
GenBank mRNA AF070614.1 - 6 - 7 1092 60
AF145713.1 - 7 - 8 1477 60
AF145714.1 - 6 - 7 781 60
AF145715.1 - 7 - 8 1438 60
AK091871.1 - 6 - 7 924 60
AK314662.1 - 7 - 8 1925 60
BC005947.1 - 6 - 7 819 60
BC036535.2 - 7 - 8 1982 60
CA431197.1 - 1 - 2 60

Gene Symbol
IQCJ-SCHIP1
Entrez Gene ID
100505385
Gene Name
IQCJ-SCHIP1 readthrough
Gene Aliases
-
Chromosome Location
Chr.3: 159069252 - 159897366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197113.1 NP_001184042.1 10 - 11 1817 60
NM_001197114.1 NP_001184043.1 9 - 10 1736 60
GenBank mRNA AK096479.1 - 10 - 11 1723 60
DQ157847.1 - 8 - 9 1555 60
DQ157848.1 - 9 - 10 1636 60

View Full Product Details