Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZIC4
Entrez Gene ID
84107
Gene Name
Zic family member 4
Gene Aliases
-
Chromosome Location
Chr.3: 147386048 - 147406893 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF332509.1 - 1 - 1 2424 114

View Full Product Details