The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080412.2 | NP_001073881.2 | 3 - 4 | 506 | 93 |
GenBank mRNA | AK311501.1 | - | 2 - 3 | 393 | 93 |
BC015444.1 | - | 3 - 4 | 506 | 93 |