The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005247257.1 | XP_005247314.1 | 1 - 2 | 67 | 86 |
XM_011512607.1 | XP_011510909.1 | 1 - 2 | 67 | 86 | |
XM_011512608.1 | XP_011510910.1 | 1 - 2 | 67 | 86 | |
XM_011512611.1 | XP_011510913.1 | 1 - 2 | 67 | 86 | |
XM_017006053.1 | XP_016861542.1 | 1 - 2 | 67 | 86 | |
XM_017006054.1 | XP_016861543.1 | 1 - 2 | 67 | 86 | |
XM_017006055.1 | XP_016861544.1 | 1 - 2 | 67 | 86 | |
XM_017006056.1 | XP_016861545.1 | 1 - 2 | 67 | 86 | |
XM_017006060.1 | XP_016861549.1 | 1 - 2 | 67 | 86 | |
XM_017006061.1 | XP_016861550.1 | 1 - 2 | 67 | 86 |